Ammonium iron(III) oxalate hydrate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Ferric ammonium oxalate

F1002-125GM F1002-2.5KG F1002-500GM F1002-12KG
75872-502EA 113.3 USD
75872-502 75873-652 75873-654 76014-452
Ammonium iron(III) oxalate hydrate
Ammonium iron(III) oxalate hydrate

Ferric Ammonium Oxalate, Granular is a water soluble product used in the manufacturing of blueprint paper and for the chemical absorption on anodized aluminum, producing a range of gold colors from light champagne hues to dark gold finishes.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: (NH₄)₃Fe(C₂O₄)₃·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00013071
CAS Number: 13268-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR