Ethyl butyrylacetate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Ethyl Butyrylacetate, Ethyl 3-oxohexanoate

E2538-100ML E2538-25ML
75872-488EA 284.9 USD
75872-488 75872-490
Ethyl butyrylacetate
Ethyl butyrylacetate

Ethyl 3-Oxohexanoate is used as a general flavoring agent in foods, including condiments and seasonings.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₈H₁₄O₃
Density: 0.989
Storage Temperature: Ambient
MDL Number: MFCD00009401
CAS Number: 3249-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR