1,1,1-Tris(hydroxymethyl)propane

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1,1,1-Trimethylolpropane, Propylidynetrimethanol, Trimethylol propane, Propylidynetris(methanol), 2-Ethyl-2-(hydroxymethyl)-1,3-propanediol

E2083-2.5KG E2083-25GM E2083-500GM E2083-12KG E2083-50KGBL
75872-476EA 181.8 USD
75872-476 75872-478 75872-480 76014-442 76014-444
1,1,1-Tris(hydroxymethyl)propane
1,1,1-Tris(hydroxymethyl)propane

2-Ethyl-2-(hydroxymethyl) 1,3-Propanediol is also known as trimethylolpropane. It is primarily consumed as a precursor to alkyd resins because it contains three hydroxy functional groups.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD00004694
CAS Number: 77-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR