(4-Methoxyphenyl)acetic acid ethyl ester

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Ethyl (4-methoxyphenyl)acetate, Ethyl 2-(4-methoxyphenyl)acetate, Homoanisic acid ethyl ester, 4-Methoxyphenylacetic acid ethyl ester, Ethyl 4-Methoxyphenylacetate

E1496-100ML E1496-25ML
75872-472EA 500.65 USD
75872-472 75872-474
(4-Methoxyphenyl)acetic acid ethyl ester
(4-Methoxyphenyl)acetic acid ethyl ester
Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040760
CAS Number: 14062-18-1
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR