Ethyl orange sodium salt

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 4-Diethylaminoazobenzene-4'-sulphonic Acid Sodium Salt

E1183-25GM E1183-5GM
75872-390EA 103.1 USD
75872-390 75872-392
Ethyl orange sodium salt
Ethyl orange sodium salt
Formula: C₁₆H₁₈N₃NaO₃S
MW: 355.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007503
CAS Number: 62758-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR