EDTA ferric monosodium salt

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Ethylenedinitrilotetra-acetic acid iron (III) sodium salt, Edetic acid iron (III) monosodium salt, Edetic acid iron (III) sodium salt, Ethylenediaminetetra-acetic acid iron (III) sodium salt, Iron (III) sodium ethylenediaminetetraacetate, Ethylenedinitrilotetra-acetic acid iron (III) monosodium salt, EDTA iron sodium, Sodium ferric ethylenediaminetetraacetate, Iron (III) monosodium ethylenediaminetetraacetate, Sodium feredetate, Ethylenediaminetetra-acetic acid iron (III) monosodium salt

E1002-100GM E1002-500GM
75872-386EA 93.51 USD
75872-386 75872-388
EDTA ferric monosodium salt
EDTA ferric monosodium salt

Ethylenediaminetetraacetic Acid, Iron (III) Sodium Salt or EDTA iron(III) sodium salt and is used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₀H₁₂FeN₂NaO₈
MW: 367.05 g/mol
Melting Pt: 80
Storage Temperature: Ambient
MDL Number: MFCD00078215
CAS Number: 15708-41-5
Merck Index: 12,04076

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR