3-(Dimethylamino)phenol

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: N,N-Dimethyl-3-hydroxyaniline, 3-Dimethylaminophenol

D2854-100GM D2854-25GM
75872-366EA 347.3 USD
75872-366 75872-368
3-(Dimethylamino)phenol
3-(Dimethylamino)phenol

3-(Dimethylamino)phenol, also known as DMAP or 4-Dimethylaminophenol, is an aromatic compound used as an antidote for cyanide poisoning and to treat hydrogen sulfide toxicity.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 265
Storage Temperature: Ambient
MDL Number: MFCD00002264
CAS Number: 99-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR