Diethyl terephthalate

Supplier: SPECTRUM CHEMICAL MFG CORP

D2596-100GM D2596-25GM
75872-362EA 246.95 USD
75872-362 75872-364
Diethyl terephthalate
Diethyl terephthalate
Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039891
CAS Number: 636-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR