Dimethylammonium chloride

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Dimethylamine hydrochloride

D2396-100GMGL D2396-2.5KGPL D2396-500GMGL D2396-12KGBL D2396-25KGBL
75872-352EA 70.55 USD
75872-352 75872-354 75872-356 76014-380 76014-394
Dimethylammonium chloride
Dimethylammonium chloride
Formula: C₂H₈ClN
MW: 81.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012477
CAS Number: 506-59-2
Merck Index: 13,03255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR