2,5-Dihydroxybenzoic acid
Supplier: Spectrum Chemical Mfg. Corp.
2,5-Dihydroxybenzoic Acid, also known as Gentisic acid, is a derivative of benzoic acid and is readily oxidized and is used as an antioxidant excipient in some pharmaceutical preparations. In the laboratory, it is used as a sample matrix in matrix-assisted laser desorption/ionization (MALDI) mass spectrometry.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₇H₆O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00002460 CAS Number: 490-79-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...