1,3-Diaminopropane

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1,3-Propanediamine, Trimethylenediamine

D2049-500ML D2049-200LTBL
75872-334EA 77.08 USD
75872-334 75872-336 76014-378
1,3-Diaminopropane
1,3-Diaminopropane

1,3-Diaminopropane, also known as Trimethylenediamine, is isomeric with 1,2-diaminopropane and used in the synthesis of heterocycles. It is a simple diamine considered soluble in polar organic solvents.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₃H₁₀N₂
MW: 74.13 g/mol
Density: 0.888
Storage Temperature: Ambient
MDL Number: MFCD00008228
CAS Number: 109-76-2
UN: 2922
ADR: 86.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR