Dimethyl methylphosphonate

Supplier: SPECTRUM CHEMICAL MFG CORP

D2040-500GM D2040-200LTBL
75875-890EA 146.05 USD
75875-890 75875-930 75875-932 76014-376
Dimethyl methylphosphonate
Dimethyl methylphosphonate

Dimethyl Methylphosphonate, also known as methylphosphonic acid dimethyl ester, is an organophosphorus compound used as a flame retardant and a preignition additive for plasticizer, gasoline, and anti-foaming agents.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₃H₉O₃P
MW: 124.08 g/mol
Density: 1.145
Storage Temperature: Ambient
MDL Number: MFCD00008349
CAS Number: 756-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR