N,O-Dimethylhydroxylamine hydrochloride

Supplier: SPECTRUM CHEMICAL MFG CORP

D2014-100GM D2014-25GM
75872-330EA 386.7 USD
75872-330 75872-332
N,O-Dimethylhydroxylamine hydrochloride
N,O-Dimethylhydroxylamine hydrochloride
Formula: C₂H₈ClNO
MW: 97.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012485
CAS Number: 6638-79-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR