Diethylammonium diethyldithiocarbamate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Diethyldithiocarbamic Acid Diethylammonium Salt, Diethylammonium diethylcarbamodithioate

D1592-100GM D1592-25GM
75872-314EA 371.75 USD
75872-314 75872-316
Diethylammonium diethyldithiocarbamate
Diethylammonium diethyldithiocarbamate
Formula: C₅H₁₁NS₂
MW: 222.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013146
CAS Number: 1518-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR