2,5-Dichlorosulfanilic acid

Supplier: Spectrum Chemical Mfg. Corp.

D1546-25GM D1546-100GM
75872-288EA 173.05 USD
75872-288 75873-650
2,5-Dichlorosulfanilic acid
2,5-Dichlorosulfanilic Acid
Formula: C₆H₅Cl₂NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00025159
CAS Number: 88-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR