Disodium glutarate
Supplier: Spectrum Chemical Mfg. Corp.
D1540-100GM
D1540-25GM
75873-646EA
314.75
USD
75873-646
75873-648
Disodium glutarate
Formula:
C₅H₈O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00070497 CAS Number: 13521-83-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...