2,5-Dibromopyridine
Supplier: SPECTRUM CHEMICAL MFG CORP
D1526-100GM
D1526-25GM
75871-842EA
523.7
USD
75871-842
75872-282
2,5-Dibromopyridine
2,5-Dibromopyridine
Formula:
C₅H₃Br₂N MW: 236.89 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006221 CAS Number: 624-28-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...