Dimethyldimyristylammonium bromide

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: N,N-Dimethyl-N-tetradecyltetradecan-1-aminium bromide, Dimethylditetradecylammonium Bromide

D1514-25GM D1514-100GM
75871-840EA 166.25 USD
75871-840 75873-632
Dimethyldimyristylammonium bromide
Dimethyldimyristylammonium bromide
Formula: C₃₀H₆₄BrN
MW: 518.75 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191429
CAS Number: 68105-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR