2,3-Dichloroanisole

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1,2-Dichloro-3-methoxybenzene

D1464-100GM D1464-25GM
75872-278EA 388 USD
75872-278 75872-280
2,3-Dichloroanisole
2,3-Dichloroanisole
Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000545
CAS Number: 1984-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR