1,4-Diformylpiperazine

Supplier: SPECTRUM CHEMICAL MFG CORP

D1440-100GM D1440-25GM
75872-270EA 270 USD
75872-270 75872-272
1,4-Diformylpiperazine
1,4-Diformylpiperazine
Formula: C₆H₁₀N₂O₂
MW: 142.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006153
CAS Number: 4164-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR