2-(Dimethylamino)ethyl acetate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Acetic Acid 2-(Dimethylamino)ethyl Ester

D1418-100ML D1418-25ML
75872-250EA 259.15 USD
75872-250 75872-252
2-(Dimethylamino)ethyl acetate
2-(Dimethylamino)ethyl acetate
Formula: C₆H₁₃NO₂
MW: 131.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041912
CAS Number: 1421-89-2
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR