Disodium adipate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Disodium Adipate, Sodium adipate, Adipic Acid Disodium Salt

D1410-100GM D1410-25GM
75872-246EA 276.8 USD
75872-246 75872-248
Disodium adipate
Disodium adipate
Formula: C₆H₁₀O₄
MW: 190.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070498
CAS Number: 7486-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR