Cresyl violet (acetate)

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Cresyl echt violet, Cresyl violet, Cresyl fast violet, 5,9-Diaminobenzo[a]phenoxazin-7-ium acetate, 9-Amino-5-imino-5H-benzo[a]phenoxazine acetate salt, Cresyl Violet acetate, Gentian violet

CR125-1GM CR125-25GM CR125-5GM
75872-202EA 71.45 USD
75872-202 75872-204 75872-206
Cresyl violet (acetate)
Cresyl violet (acetate)
Formula: C₁₈H₁₅N₃O₃
MW: 321.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013151
CAS Number: 10510-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR