Choline bromide
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
2-Hydroxy-N,N,N-trimethylethan-1-aminium bromide
C3808-100GM
C3808-25GM
75872-184EA
308
USD
75872-184
75872-186
Choline bromide
Choline bromide
Formula:
C₅H₁₄BrNO MW: 184.748 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00054244 CAS Number: 1927-06-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...