Cholesterol pelargonate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Cholesterol Nonanoate, Nonanoic Acid Cholesterol Ester, Pelargonic Acid Cholesterol Ester

C3478-100GM C3478-25GM
75873-816EA 276.8 USD
75873-816 75873-818
Cholesterol pelargonate
Cholesterol pelargonate
Formula: C₃₆H₆₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00003643
CAS Number: 1182-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR