L-Cystine disodium salt monohydrate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (H-Cys-OH)2.2Na.H2O

C3035-100GM C3035-1KG C3035-25GM
75873-798EA 420.55 USD
75873-798 75873-800 75873-820
L-Cystine disodium salt monohydrate
L-Cystine disodium salt monohydrate

L-Cystine Disodium Salt, Monohydrate, also known simply as Cystine Sodium, is an amino acid and organosulfur compound which is formed by the oxidation of two cysteine molecules that covalently link by a disulfide bond.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₁₀N₂Na₂O₄S₂·H₂O
MW: 302.27 g/mol (284.26 anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD00243291
CAS Number: 199329-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR