2-Chloro-1-methylpyridinium iodide

Supplier: SPECTRUM CHEMICAL MFG CORP

C2899-100GM C2899-25GM
75872-162EA 284.9 USD
75872-162 75872-164
2-Chloro-1-methylpyridinium iodide
2-Chloro-1-methylpyridinium iodide
Formula: C₆H₇ClIN
MW: 255.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011984
CAS Number: 14338-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR