4-Chlorobutyryl chloride
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
4-Chlorobutanoyl Chloride
C2226-100ML
C2226-500ML
75872-148EA
124.4
USD
75872-148
75872-150
4-Chlorobutyryl chloride
4-Chlorobutyryl chloride
Formula:
C₄H₆Cl₂O MW: 141 g/mol Density: 1.258 Storage Temperature: Ambient |
MDL Number:
MFCD00000754 CAS Number: 4635-59-0 UN: 2927 ADR: 6.18,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...