Cyclohexylamine hydrochloride
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Cyclohexylamine Hydrochloride
C2127-100GM
C2127-25GM
C2127-500GM
75872-138EA
147.25
USD
75872-138
75872-140
75872-142
Cyclohexylamine hydrochloride
Cyclohexylamine hydrochloride
Cyclohexylamine Hydrochloride is an organic compound that is an aliphatic amine. The compound is a high-melting solid. Cyclohexylamine hydrochloride is used as an intermediate in the synthesis of other organic compounds.
Formula:
C₆H₁₃N MW: 135.64 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00050540 CAS Number: 4998-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...