Choline hydrogen tartrate USP dietary supplement
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
2-Hydroxy-N,N,N-trimethylethan-1-aminium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Choline acid tartrate, CBT, 2-(Hydroxyethyl)trimethylammonium bitartrate, Choline bitartrate
C2052-125GM
C2052-12KG
C2052-2.5KG
C2052-500GM
C2052-50KGBL
76014-314EA
89.31
USD
76014-314
76014-316
76014-318
76014-320
76014-322
Choline hydrogen tartrate USP dietary supplement
Choline hydrogen tartrate
Choline Bitartrate, USP Dietary Supplement.
All Spectrum Chemical USP Dietary Supplement grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formula:
C₉H₁₉NO₇ MW: 253.25 g/mol Melting Pt: 151…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036332 CAS Number: 87-67-2 |
Specification Test Results
APPEARANCE | |
IDENTIFICATION (A) FTIR | SPECTRUM MATCHES REFERENCE |
IDENTIFICATION (B) | WHITE PRECIPITATE FORMS |
ASSAY (ANHYDROUS BASIS) | 99.0-100.5 % |
OPTICAL ROTATION | +17.5° to +18.5° |
pH (1 in 10 SOLUTION) | 3.0 - 4.0 |
ELEMENTAL IMPURITIES | AS REPORTED |
ARSENIC | min. 2 ppm |
LEAD | min. 0.3 ppm |
TOTAL AMINES | min. 10 µg/g |
1,4-DIOXANE | min. 10 µg/g |
RESIDUE ON IGNITION | min. 0.1 % |
WATER DETERMINATION | min. 0.5 % |
CHROMATOGRAPHIC PURITY: | min. |
INDIVIDUAL IMPURITIES | min. 0.3 % |
TOTAL IMPURITIES | min. 2.0 % |
RESIDUAL SOLVENTS | AS REPORTED |
CLASS 3 (solvent) / ETHANOL | . |
MONOGRAPH EDITION |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...