18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane)

Supplier: Spectrum Chemical Mfg. Corp.

C1799-100GM C1799-250GM C1799-25GM
75872-128EA 355.45 USD
75872-128 75872-130 75872-132
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane)
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane)
Formula: C₁₂H₂₄O₆
Storage Temperature: Ambient
MDL Number: MFCD00005113
CAS Number: 17455-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR