L(-)-Carnitine free base USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate

C1399-100GM C1399-1KG C1399-25GM C1399-25KGBL C1399-500GM C1399-5KGBL
76014-278EA 533.85 USD
76014-278 76014-280 76014-282 76014-284 76014-286 76014-288
L(-)-Carnitine free base USP
L(-)-Carnitine

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₇H₁₅NO₃
MW: 161.20 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00038747
CAS Number: 541-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR