4-Bromobenzenesulfonyl chloride

Supplier: SPECTRUM CHEMICAL MFG CORP

B2904-100GM B2904-25GM
75872-064EA 336.45 USD
75872-064 75872-066
4-Bromobenzenesulfonyl chloride
4-Bromobenzenesulphonyl chloride
Formula: C₆H₄BrClO₂S
MW: 255.52 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007437
CAS Number: 98-58-8
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR