N-(3-Bromopropyl)phthalimide

Supplier: SPECTRUM CHEMICAL MFG CORP

B2457-100GM B2457-25GM
75872-054EA 416.55 USD
75872-054 75872-056
N-(3-Bromopropyl)phthalimide
N-(3-Bromopropyl)phthalimide
Formula: C₁₁H₁₀BrNO₂
MW: 268.11 g/mol
Boiling Pt: 433
Storage Temperature: Ambient
MDL Number: MFCD00005904
CAS Number: 5460-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR