Bis(4-fluorophenyl) sulfone

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 4-Fluorophenyl sulphone, 4,4'-Difluoro diphenyl sulphone, 1-Fluor-4-((4-fluorphenyl)sulphonyl)benzol, 4,4'-sulphonylbis(fluorobenzene)

B1996-100GM B1996-500GM B1996-25KGBL
75872-022EA 386.7 USD
75872-022 75872-024 76014-158
Bis(4-fluorophenyl) sulfone
Bis(4-fluorophenyl)sulphone
Formula: C₁₂H₈F₂O₂S
MW: 254.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000350
CAS Number: 383-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR