N-Benzylacetamide

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Lacosamide EP Impurity G, N-Acetylbenzylamine

B1536-100GM B1536-25GM
75872-006EA 495.25 USD
75872-006 75872-008
N-Benzylacetamide
N-Benzylacetamide
Formula: C₉H₁₁NO
MW: 149.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059204
CAS Number: 588-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR