Benzyl methyl ether

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (Methoxymethyl)benzene

B1486-100ML B1486-25ML
75871-980EA 275.4 USD
75871-980 75871-982
Benzyl methyl ether
Benzyl methyl ether
Formula: C₈H₁₀O
MW: 122.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00025901
CAS Number: 538-86-3
UN: 3271
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR