4'-Butylacetophenone

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 4'-n-Butylacetophenone, 4-Butylacetophenone, p-Butylacetophenone

B1458-100GM B1458-25GM
75871-954EA 489.75 USD
75871-954 75871-956
4'-Butylacetophenone
4'-Butylacetophenone
Formula: C₁₂H₁₆O
MW: 176.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017500
CAS Number: 37920-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR