4-Bromophenyl phenyl ether (BDE No. 3)

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 4-Bromodiphenyl ether, 1-Bromo-4-phenoxybenzene

B1456-100GM B1456-25GM
75871-950EA 287.65 USD
75871-950 75871-952
4-Bromophenyl phenyl ether (BDE No. 3)
4-Bromophenyl phenyl ether (BDE No. 3)
Formula: C₁₂H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD00000094
CAS Number: 101-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR