4-(2-Benzoxazolyl)-4'-(5-methyl-2-benzoxazolyl)stilbene

Supplier: SPECTRUM CHEMICAL MFG CORP

B1454-100GM B1454-25GM
75871-812EA 282.15 USD
75871-812 75871-814
4-(2-Benzoxazolyl)-4'-(5-methyl-2-benzoxazolyl)stilbene
4-(2-Benzoxazolyl)-4'-(5-methyl-2-benzoxazolyl)stilbene
Formula: C₂₉H₂₀N₂O₂
MW: 428.49 g/mol
Storage Temperature: Ambient
CAS Number: 5242-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR