3-Buten-1-ol

Supplier: Spectrum Chemical Mfg. Corp.

B1452-25ML B1452-100ML
75871-810EA 144.55 USD
75871-810 75871-948
3-Buten-1-ol
3-Buten-1-ol
Formula: C₄H₈O
MDL Number: MFCD00002959
CAS Number: 627-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR