1-Benzyl-1-phenylhydrazine hydrochloride

Supplier: Spectrum Chemical Mfg. Corp.

B1424-100GM B1424-25GM
75871-924EA 293.05 USD
75871-924 75871-926
1-Benzyl-1-phenylhydrazine hydrochloride
1-Benzyl-1-phenylhydrazine hydrochloride
Formula: C₁₃H₁₅ClN₂
Storage Temperature: Ambient
MDL Number: MFCD00050690
CAS Number: 5705-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR