L(+)-Aspartic acid

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (S)-(+)-Aminosuccinic acid (S)-Aminobutanedioic acid (S)-Aminosuccinic acid H-Asp-OH L-Asparaginic acid L(+)-Aspartic acid, (S)-(+)-Aspartic acid, L(+)-2-Amino-1,4-butanedioic acid, (S)-(+)-2-Amino-1,4-butanedioic acid, L-Aspartic acid, H-Asp-OH, L-α-Aminosuccinic acid, L-αlpha-Aminosuccinic acid, (S)-2-Aminosuccinic acid, Asp, D

AS125-100GM AS125-1KG
75871-904EA 74.6 USD
75871-904 75871-906
L(+)-Aspartic acid
L(+)-Aspartic acid

L-Aspartic Acid is a nonessential amino acid. Aspartic acid is used as a component of parenteral and enteral nutrition. In pharmaceutical agents aspartic acid is used as an ammoniac detoxicating agent, hepar function accelerator and fatigue refresher. Aspartic acid is also widely utilized for mineral supplementation in the form of its potassium, magnesium or calcium salts.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄H₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00002616
CAS Number: 56-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR