β-Alanine
Supplier: Spectrum Chemical Mfg. Corp.
AL120-100GM
AL120-2.5KG
AL120-500GM
AL120-12KG
AL120-50KGBL
75871-886EA
109.27
USD
75871-886
75871-888
75871-890
76014-196
76014-198
β-Alanine
β-Alanine
Formula:
C₃H₇NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00008200 CAS Number: 107-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...