Adenosine
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
Adenosine is a purine nucleotide with a molecule of adenine bound to a ribose sugar molecule. It has anti-inflammatory properties as well as inhibitory effects on the central nervous system.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₀H₁₃N₅O₄ MW: 267.24 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00005752 CAS Number: 58-61-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...