2-Phenylaniline

Supplier: SPECTRUM CHEMICAL MFG CORP

A2776-100GM A2776-25GM
75871-846EA 299.8 USD
75871-846 75871-848
2-Phenylaniline
2-Phenylaniline
Formula: C₁₂H₁₁N
MW: 169.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007701
CAS Number: 90-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR