m-Anisaldehyde

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 3-Methoxybenzaldehyde

A2346-100ML A2346-25ML
75871-794EA 283.55 USD
75871-794 75871-796
m-Anisaldehyde
m-Anisaldehyde
Formula: C₈H₈O₂
MW: 136.15 g/mol
Density: 1.119
Storage Temperature: Ambient
MDL Number: MFCD00003361
CAS Number: 591-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR