Allyl cyclohexanepropionate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Allyl Cyclohexanepropionate, Cyclohexanepropanoic acid 2-propen-1-yl ester, Allyl 3-cyclohexylpropanoate, 2-Propenyl 3-cyclohexylpropanoate, Cyclohexanepropionic acid allyl ester, Allyl 3-cyclohexylpropionate, 3-Cyclohexylpropionic acid Allyl Ester Cyclohexanepropionic acid Allyl Ester

A2315-100ML A2315-25ML
75871-790EA 423.3 USD
75871-790 75871-792
Allyl cyclohexanepropionate
Allyl cyclohexanepropionate
Formula: C₁₂H₂₀O₂
MW: 196.29 g/mol
Density: 0.948
Storage Temperature: Ambient
MDL Number: MFCD00059557
CAS Number: 2705-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR