Triammonium citrate

Supplier: Spectrum Chemical Mfg. Corp.

A2040-100GM A2040-2.5KG A2040-500GM A2040-12KGBL A2040-50KGBL
75871-722EA 76.7 USD
75871-722 75871-724 75871-726 76014-162 76014-164
Triammonium citrate
Triammonium citrate
Formula: C₆H₁₇N₃O₇
Storage Temperature: Ambient
MDL Number: MFCD00036406
CAS Number: 3458-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR