Ethyl hydrogen adipate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Adipic acid monoethyl ester, Adipic acid ethyl ester, 6-Ethoxy-6-oxohexanoic acid, 1,6-Hexanedioic acid monoethyl ester, Monoethyl adipate, 1,6-Hexanedioic acid ethyl ester

A2023-100GM A2023-25GM
75871-806EA 291.7 USD
75871-806 75871-808
Ethyl hydrogen adipate
Ethyl hydrogen adipate
Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Boiling Pt: 286
Melting Pt: 27.4
Density: 0.98
Storage Temperature: Ambient
MDL Number: MFCD00004419
CAS Number: 626-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR